accessibility menu, dialog, popup

UserName

BrO

CAS RN 32752-54-8

IUPAC Name: [4-(bromomethyl)phenyl](phenyl)methanone
Synonyms: [4-(bromomethyl)phenyl](phenyl)methanone
Molecular Weight (g/mol): 275.15
Molecular Formula: C14H11BrO
InChi Key: RYULULVJWLRDQH-UHFFFAOYSA-N
SMILES: BrCC1=CC=C(C=C1)C(=O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results