missing translation for 'onlineSavingsMsg'
Learn More

4-(Bromomethyl)benzenesulfonyl Chloride 95.0+%, TCI America™

Catalog No. B39365G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39365G 5 g
B39361G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39365G Supplier TCI America Supplier No. B39365G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 66176-39-4
Molecular Formula C7H6BrClO2S
Molecular Weight (g/mol) 269.537
MDL Number MFCD00156129
InChI Key QXTQWYZHHMQSQH-UHFFFAOYSA-N
Synonym 4-bromomethyl benzenesulfonyl chloride, 4-bromomethyl benzene-1-sulfonyl chloride, 4-bromomethyl benzenesulfonylchloride, alpha-bromo-p-toluenesulphonyl chloride, alpha-bromo-p-toluenesulfonyl chloride, 4-bromomethyl benzenesulphonyl chloride, 4-bromomethyl benzene-1-sulfonylchloride, buttpark 94\04-08, 4-bromomethyl phenyl chlorosulfone, 4-bromomethylene benzenesulfonyl chloride
PubChem CID 2734409
IUPAC Name 4-(bromomethyl)benzenesulfonyl chloride
SMILES C1=CC(=CC=C1CBr)S(=O)(=O)Cl

Specifications

Melting Point 75°C
Color White-Yellow
Quantity 5 g
UN Number 3261
Formula Weight 269.54
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Bromomethyl)benzenesulfonyl Chloride