accessibility menu, dialog, popup

UserName

CAS RN 66176-39-4

IUPAC Name: 4-(bromomethyl)benzene-1-sulfonyl chloride
Synonyms: 4-(bromomethyl)benzenesulfonyl chloride
Molecular Weight (g/mol): 269.54
Molecular Formula: C7H6BrClO2S
InChi Key: QXTQWYZHHMQSQH-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC=C(CBr)C=C1
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (4)
  • (3)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (4)