missing translation for 'onlineSavingsMsg'
Learn More

4-(Bromomethyl)benzenesulfonyl chloride, 95%

Catalog No. p-7080556
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1012006 5 g
AAA1012014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1012006 Supplier Thermo Scientific Chemicals Supplier No. A1012006
Only null left

CAS: 66176-39-4 | C7H6BrClO2S | 269.537 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 66176-39-4
Molecular Formula C7H6BrClO2S
Molecular Weight (g/mol) 269.537
MDL Number MFCD00156129
InChI Key QXTQWYZHHMQSQH-UHFFFAOYSA-N
Synonym 4-bromomethyl benzenesulfonyl chloride, 4-bromomethyl benzene-1-sulfonyl chloride, 4-bromomethyl benzenesulfonylchloride, alpha-bromo-p-toluenesulphonyl chloride, alpha-bromo-p-toluenesulfonyl chloride, 4-bromomethyl benzenesulphonyl chloride, 4-bromomethyl benzene-1-sulfonylchloride, buttpark 94\04-08, 4-bromomethyl phenyl chlorosulfone, 4-bromomethylene benzenesulfonyl chloride
PubChem CID 2734409
IUPAC Name 4-(bromomethyl)benzenesulfonyl chloride
SMILES C1=CC(=CC=C1CBr)S(=O)(=O)Cl

Specifications

Melting Point 72°C to 75°C
Quantity 5 g
UN Number UN3261
Beilstein 2364328
Sensitivity Moisture sensitive
Formula Weight 269.55
Percent Purity 95%
Chemical Name or Material 4- (Bromomethyl)benzenesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only