missing translation for 'onlineSavingsMsg'
Learn More

4-Bromomethyl-3-fluorobenzonitrile, 95%, Thermo Scientific Chemicals

Catalog No. p-7054825
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6324503 1 g
AAH6324506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6324503 Supplier Thermo Scientific Chemicals Supplier No. H6324503
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 105942-09-4
Molecular Formula C8H5BrFN
Molecular Weight (g/mol) 214.04
MDL Number MFCD07368341
InChI Key ZESZAIOGACKOMB-UHFFFAOYSA-N
Synonym 4-bromomethyl-3-fluorobenzonitrile, 4-cyano-2-fluorobenzyl bromide, 2-fluoro-4-cyanobenzyl bromide, 2-fluoro-4-cyanbenzyl bromide, benzonitrile, 4-bromomethyl-3-fluoro, 2-fluoro-4-cyanobenzylbromide, 3-fluoro-4-bromomethyl benzonitrile, 4-bromomethyl-3-fluoro-benzonitrile, 4-bromomethyl-3-fluorobenzenecarbonitrile, pubchem4907
PubChem CID 2783149
IUPAC Name 4-(bromomethyl)-3-fluorobenzonitrile
SMILES FC1=C(CBr)C=CC(=C1)C#N

Specifications

Melting Point 75°C to 79°C (decomposition)
Color White
Quantity 1 g
UN Number UN2923
Beilstein 7758134
Formula Weight 214.04
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 4-Bromomethyl-3-fluorobenzonitrile
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only