missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Bromomethyl-2-cyanobiphenyl 98.0+%, TCI America™
Supplier: TCI America B180625G
Specifications
| 4′-Bromomethyl-2-cyanobiphenyl | |
| 127°C | |
| C14H10BrN | |
| 25 g | |
| LFFIEVAMVPCZNA-UHFFFAOYSA-N | |
| 2-[4-(bromomethyl)phenyl]benzonitrile | |
| 1501912 | |
| ≥98.0% (GC) |
| 114772-54-2 | |
| White | |
| MFCD00671503 | |
| 4'-bromomethyl-2-cyanobiphenyl, 2-4-bromomethyl phenyl benzonitrile, 2-cyano-4'-bromomethylbiphenyl, 4'-bromomethyl-1,1'-biphenyl-2-carbonitrile, 2-4-bromo-methylphenyl benzonitrile, 4'-bromomethyl-2-biphenylcarbonitrile, 4'-bromomethylbiphenyl-2-carbonitrile, 2'-cyano-4-bromomethylbiphenyl, 4-2-cyanophenyl benzyl bromide, unii-ui5jr3k10f | |
| C1=CC=C(C(=C1)C#N)C2=CC=C(C=C2)CBr | |
| 272.145 | |
| 272.15 | |
| Crystalline Powder |
Chemical Identifiers
| 114772-54-2 | |
| 272.145 | |
| LFFIEVAMVPCZNA-UHFFFAOYSA-N | |
| 1501912 | |
| C1=CC=C(C(=C1)C#N)C2=CC=C(C=C2)CBr |
| C14H10BrN | |
| MFCD00671503 | |
| 4'-bromomethyl-2-cyanobiphenyl, 2-4-bromomethyl phenyl benzonitrile, 2-cyano-4'-bromomethylbiphenyl, 4'-bromomethyl-1,1'-biphenyl-2-carbonitrile, 2-4-bromo-methylphenyl benzonitrile, 4'-bromomethyl-2-biphenylcarbonitrile, 4'-bromomethylbiphenyl-2-carbonitrile, 2'-cyano-4-bromomethylbiphenyl, 4-2-cyanophenyl benzyl bromide, unii-ui5jr3k10f | |
| 2-[4-(bromomethyl)phenyl]benzonitrile |
Safety and Handling
TSCA : No