accessibility menu, dialog, popup

UserName

CAS RN 114772-54-2

IUPAC Name: 4'-(bromomethyl)-[1,1'-biphenyl]-2-carbonitrile
Synonyms: 4'-(bromomethyl)-[1,1'-biphenyl]-2-carbonitrile
Molecular Weight (g/mol): 272.15
Molecular Formula: C14H10BrN
InChi Key: LFFIEVAMVPCZNA-UHFFFAOYSA-N
SMILES: BrCC1=CC=C(C=C1)C1=CC=CC=C1C#N
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)