missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromomethyl-2-cyanobiphenyl 98.0+%, TCI America™

Catalog No. B18065G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18065G 5 g
B180625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B18065G Supplier TCI America Supplier No. B18065G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 114772-54-2
Molecular Formula C14H10BrN
Molecular Weight (g/mol) 272.145
MDL Number MFCD00671503
InChI Key LFFIEVAMVPCZNA-UHFFFAOYSA-N
Synonym 4'-bromomethyl-2-cyanobiphenyl, 2-4-bromomethyl phenyl benzonitrile, 2-cyano-4'-bromomethylbiphenyl, 4'-bromomethyl-1,1'-biphenyl-2-carbonitrile, 2-4-bromo-methylphenyl benzonitrile, 4'-bromomethyl-2-biphenylcarbonitrile, 4'-bromomethylbiphenyl-2-carbonitrile, 2'-cyano-4-bromomethylbiphenyl, 4-2-cyanophenyl benzyl bromide, unii-ui5jr3k10f
PubChem CID 1501912
IUPAC Name 2-[4-(bromomethyl)phenyl]benzonitrile
SMILES C1=CC=C(C(=C1)C#N)C2=CC=C(C=C2)CBr

Specifications

Melting Point 127°C
Color White
Quantity 5 g
Formula Weight 272.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Bromomethyl-2-cyanobiphenyl