Learn More
(4-Bromobutoxy)benzene, 97%
CAS: 1200-03-9 | C10H13BrO | 229.12 g/mol
Supplier: Thermo Scientific Chemicals A1495614
Description
(4-Bromobutoxy)benzene is used to produce 5-phenoxy-valeric acid with carbon dioxide and reagent Mg.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(4-Bromobutoxy)benzene is used to produce 5-phenoxy-valeric acid with carbon dioxide and reagent Mg.
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
| (4-Bromobutoxy)benzene | |
| 40°C to 43°C | |
| >110°C (230°F) | |
| C10H13BrO | |
| 25 g | |
| 4-bromobutoxy benzene, 4-phenoxybutyl bromide, 4-bromobutyl phenyl ether, benzene, 4-bromobutoxy, 1-bromo-4-phenoxybutane, phenoxybutyl bromide, 4-phenoxybutylbromide, benzene, bromobutoxy, 4-bromo-1-phenoxybutane, 4-brombutoxy benzol | |
| QBLISOIWPZSVIK-UHFFFAOYSA-N | |
| 4-bromobutoxybenzene | |
| 70986 | |
| 97% |
| 1200-03-9 | |
| 153°C to 154°C (18 mmHg) | |
| Odorless | |
| MFCD00000262 | |
| 2044374 | |
| Insoluble in water. | |
| BrCCCCOC1=CC=CC=C1 | |
| 229.12 | |
| 229.12 |
Chemical Identifiers
| 1200-03-9 | |
| 229.12 | |
| QBLISOIWPZSVIK-UHFFFAOYSA-N | |
| 70986 | |
| BrCCCCOC1=CC=CC=C1 |
| C10H13BrO | |
| MFCD00000262 | |
| 4-bromobutoxy benzene, 4-phenoxybutyl bromide, 4-bromobutyl phenyl ether, benzene, 4-bromobutoxy, 1-bromo-4-phenoxybutane, phenoxybutyl bromide, 4-phenoxybutylbromide, benzene, bromobutoxy, 4-bromo-1-phenoxybutane, 4-brombutoxy benzol | |
| 4-bromobutoxybenzene |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 214-849-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only