missing translation for 'onlineSavingsMsg'
Learn More

(4-Bromobutoxy)benzene, 97%

Catalog No. p-7022266
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1495606 5 g
AAA1495614 25 g
AAA1495622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1495606 Supplier Thermo Scientific Chemicals Supplier No. A1495606

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1200-03-9 | C10H13BrO | 229.12 g/mol

(4-Bromobutoxy)benzene is used to produce 5-phenoxy-valeric acid with carbon dioxide and reagent Mg.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(4-Bromobutoxy)benzene is used to produce 5-phenoxy-valeric acid with carbon dioxide and reagent Mg.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 1200-03-9
Molecular Formula C10H13BrO
Molecular Weight (g/mol) 229.12
MDL Number MFCD00000262
InChI Key QBLISOIWPZSVIK-UHFFFAOYSA-N
Synonym 4-bromobutoxy benzene, 4-phenoxybutyl bromide, 4-bromobutyl phenyl ether, benzene, 4-bromobutoxy, 1-bromo-4-phenoxybutane, phenoxybutyl bromide, 4-phenoxybutylbromide, benzene, bromobutoxy, 4-bromo-1-phenoxybutane, 4-brombutoxy benzol
PubChem CID 70986
IUPAC Name 4-bromobutoxybenzene
SMILES BrCCCCOC1=CC=CC=C1

Specifications

Melting Point 40°C to 43°C
Boiling Point 153°C to 154°C (18 mmHg)
Flash Point >110°C (230°F)
Odor Odorless
Quantity 5 g
Beilstein 2044374
Solubility Information Insoluble in water.
Formula Weight 229.12
Percent Purity 97%
Chemical Name or Material (4-Bromobutoxy)benzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-849-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only