missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzyl Chloride 98.0+%, TCI America™

Catalog No. p-7119910
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30425G 5 g
B304225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30425G Supplier TCI America Supplier No. B30425G

Chemical Identifiers

CAS 589-17-3
Molecular Formula C7H6BrCl
Molecular Weight (g/mol) 205.48
MDL Number MFCD00040867
InChI Key BSIIGUGKOPPTPZ-UHFFFAOYSA-N
Synonym 1-bromo-4-chloromethyl benzene, 4-bromobenzyl chloride, p-bromobenzyl chloride, 4-bromobenzylchloride, 4-bromo-alpha-chlorotoluene, benzene, 1-bromo-4-chloromethyl, ccris 5105, p-bromo-alpha-chlorotoluene, pubchem2478, 4-bromo-benzylchloride
PubChem CID 68528
IUPAC Name 1-bromo-4-(chloromethyl)benzene
SMILES ClCC1=CC=C(Br)C=C1

Specifications

Melting Point 39°C
Color White-Yellow
Boiling Point 236°C
Quantity 5 g
UN Number 3261
Formula Weight 205.48
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromobenzyl Chloride