accessibility menu, dialog, popup

UserName

ClBr

CAS RN 589-17-3

IUPAC Name: 1-bromo-4-(chloromethyl)benzene
Synonyms: p-bromobenzyl chloride
Molecular Weight (g/mol): 205.48
Molecular Formula: C7H6BrCl
InChi Key: BSIIGUGKOPPTPZ-UHFFFAOYSA-N
SMILES: ClCC1=CC=C(Br)C=C1
Boiling Point 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)