missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzyl Bromide 98.0+%, TCI America™

Catalog No. B1002500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1002500G 500 g
B100225G 25 g
B1002100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B1002500G Supplier TCI America Supplier No. B1002500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 589-15-1
Molecular Formula C7H6Br2
Molecular Weight (g/mol) 249.933
MDL Number MFCD00000179
InChI Key YLRBJYMANQKEAW-UHFFFAOYSA-N
Synonym 4-bromobenzyl bromide, 1-bromo-4-bromomethyl benzene, p-bromobenzyl bromide, alpha,p-dibromotoluene, alpha,4-dibromotoluene, benzene, 1-bromo-4-bromomethyl, 4-bromobenzylbromide, unii-3d1ttl8bgn, a,p-dibromotoluene, 4-bromo benzyl bromide
PubChem CID 68527
IUPAC Name 1-bromo-4-(bromomethyl)benzene
SMILES C1=CC(=CC=C1CBr)Br

Specifications

Melting Point 61°C
Color White
Boiling Point 124°C
Quantity 500 g
UN Number 3261
Formula Weight 249.93
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromobenzyl Bromide
EINECSNumber (3)-4330
TSCA Yes