accessibility menu, dialog, popup

UserName

BrBr

CAS RN 589-15-1

IUPAC Name: 1-bromo-4-(bromomethyl)benzene
Synonyms: 1-bromo-4-(bromomethyl)benzene
Molecular Weight (g/mol): 249.93
Molecular Formula: C7H6Br2
InChi Key: YLRBJYMANQKEAW-UHFFFAOYSA-N
SMILES: BrCC1=CC=C(Br)C=C1
Boiling Point 
  • (2)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (3)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)