Learn More
4-Bromobenzyl bromide, 98+%
CAS: 589-15-1 | C7H6Br2 | 249.933 g/mol
$90.92 - $218.90
Chemical Identifiers
| CAS | 589-15-1 |
|---|---|
| Molecular Formula | C7H6Br2 |
| Molecular Weight (g/mol) | 249.933 |
| MDL Number | MFCD00000179 |
| InChI Key | YLRBJYMANQKEAW-UHFFFAOYSA-N |
| Synonym | 4-bromobenzyl bromide, 1-bromo-4-bromomethyl benzene, p-bromobenzyl bromide, alpha,p-dibromotoluene, alpha,4-dibromotoluene, benzene, 1-bromo-4-bromomethyl, 4-bromobenzylbromide, unii-3d1ttl8bgn, a,p-dibromotoluene, 4-bromo benzyl bromide |
| PubChem CID | 68527 |
| IUPAC Name | 1-bromo-4-(bromomethyl)benzene |
| SMILES | C1=CC(=CC=C1CBr)Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1089214
|
Thermo Scientific Chemicals
A1089214 |
25 g |
Each for $90.92
|
|
|||||
|
AAA1089222
|
Thermo Scientific Chemicals
A1089222 |
100 g |
Each for $218.90
|
|
|||||
Description
4-Bromobenzyl bromide is used to prepare a 20-member aminomethyl-substituted biaryl library via sequential N-alkylation of various amines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Bromobenzyl bromide is used to prepare a 20-member aminomethyl-substituted biaryl library via sequential N-alkylation of various amines.
Solubility
Easily soluble in hot alcohol, ether, carbon disulfide, benzene and glacial acetic acid. Soluble in water and alcohol.
Notes
Keep containers tightly closed in a dry, cool and well-ventilated place. Store at 4°C. Incompatible with strong oxidizing agents. It is sensitive to light.
Chemical Identifiers
| 589-15-1 | |
| 249.933 | |
| YLRBJYMANQKEAW-UHFFFAOYSA-N | |
| 68527 | |
| C1=CC(=CC=C1CBr)Br |
| C7H6Br2 | |
| MFCD00000179 | |
| 4-bromobenzyl bromide, 1-bromo-4-bromomethyl benzene, p-bromobenzyl bromide, alpha,p-dibromotoluene, alpha,4-dibromotoluene, benzene, 1-bromo-4-bromomethyl, 4-bromobenzylbromide, unii-3d1ttl8bgn, a,p-dibromotoluene, 4-bromo benzyl bromide | |
| 1-bromo-4-(bromomethyl)benzene |
Specifications
| 589-15-1 | |
| 115°C to 124°C (12 mmHg) | |
| MFCD00000179 | |
| UN1759 | |
| 14,1409 | |
| Easily soluble in hot alcohol,ether,carbon disulfide,benzene and glacial acetic acid. Soluble in water and alcohol. | |
| C1=CC(=CC=C1CBr)Br | |
| 249.933 | |
| 249.94 | |
| 4-Bromobenzyl bromide |
| 60°C to 64°C | |
| C7H6Br2 | |
| 25 g | |
| 606498 | |
| 4-bromobenzyl bromide, 1-bromo-4-bromomethyl benzene, p-bromobenzyl bromide, alpha,p-dibromotoluene, alpha,4-dibromotoluene, benzene, 1-bromo-4-bromomethyl, 4-bromobenzylbromide, unii-3d1ttl8bgn, a,p-dibromotoluene, 4-bromo benzyl bromide | |
| YLRBJYMANQKEAW-UHFFFAOYSA-N | |
| 1-bromo-4-(bromomethyl)benzene | |
| 68527 | |
| ≥98% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
EINECSNumber : 209-636-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only