missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzyl bromide, 98+%

Catalog No. AAA1089214
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1089214 25 g
AAA1089222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1089214 Supplier Thermo Scientific Chemicals Supplier No. A1089214

CAS: 589-15-1 | C7H6Br2 | 249.933 g/mol

4-Bromobenzyl bromide is used to prepare a 20-member aminomethyl-substituted biaryl library via sequential N-alkylation of various amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobenzyl bromide is used to prepare a 20-member aminomethyl-substituted biaryl library via sequential N-alkylation of various amines.

Solubility
Easily soluble in hot alcohol, ether, carbon disulfide, benzene and glacial acetic acid. Soluble in water and alcohol.

Notes
Keep containers tightly closed in a dry, cool and well-ventilated place. Store at 4°C. Incompatible with strong oxidizing agents. It is sensitive to light.

Chemical Identifiers

CAS 589-15-1
Molecular Formula C7H6Br2
Molecular Weight (g/mol) 249.933
MDL Number MFCD00000179
InChI Key YLRBJYMANQKEAW-UHFFFAOYSA-N
Synonym 4-bromobenzyl bromide, 1-bromo-4-bromomethyl benzene, p-bromobenzyl bromide, alpha,p-dibromotoluene, alpha,4-dibromotoluene, benzene, 1-bromo-4-bromomethyl, 4-bromobenzylbromide, unii-3d1ttl8bgn, a,p-dibromotoluene, 4-bromo benzyl bromide
PubChem CID 68527
IUPAC Name 1-bromo-4-(bromomethyl)benzene
SMILES C1=CC(=CC=C1CBr)Br

Specifications

Melting Point 60°C to 64°C
Boiling Point 115°C to 124°C (12 mmHg)
Quantity 25 g
UN Number UN1759
Beilstein 606498
Merck Index 14,1409
Solubility Information Easily soluble in hot alcohol,ether,carbon disulfide,benzene and glacial acetic acid. Soluble in water and alcohol.
Formula Weight 249.94
Percent Purity ≥98%
Chemical Name or Material 4-Bromobenzyl bromide
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
EINECSNumber 209-636-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only