Learn More
4-Bromobenzyl bromide, 98%
CAS: 589-15-1 | C7H6Br2 | 249.93 g/mol
$91.20 - $91.20
Chemical Identifiers
| CAS | 589-15-1 |
|---|---|
| Molecular Formula | C7H6Br2 |
| Molecular Weight (g/mol) | 249.93 |
| MDL Number | MFCD00000179 |
| InChI Key | YLRBJYMANQKEAW-UHFFFAOYSA-N |
| Synonym | 4-bromobenzyl bromide, 1-bromo-4-bromomethyl benzene, p-bromobenzyl bromide, alpha,p-dibromotoluene, alpha,4-dibromotoluene, benzene, 1-bromo-4-bromomethyl, 4-bromobenzylbromide, unii-3d1ttl8bgn, a,p-dibromotoluene, 4-bromo benzyl bromide |
| PubChem CID | 68527 |
| IUPAC Name | 1-bromo-4-(bromomethyl)benzene |
| SMILES | C1=CC(=CC=C1CBr)Br |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC150410250
|
Thermo Scientific Chemicals
150410250 |
25 g | Glass bottle |
Each for $91.20
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 589-15-1 | |
| 249.93 | |
| YLRBJYMANQKEAW-UHFFFAOYSA-N | |
| 68527 | |
| C1=CC(=CC=C1CBr)Br |
| C7H6Br2 | |
| MFCD00000179 | |
| 4-bromobenzyl bromide, 1-bromo-4-bromomethyl benzene, p-bromobenzyl bromide, alpha,p-dibromotoluene, alpha,4-dibromotoluene, benzene, 1-bromo-4-bromomethyl, 4-bromobenzylbromide, unii-3d1ttl8bgn, a,p-dibromotoluene, 4-bromo benzyl bromide | |
| 1-bromo-4-(bromomethyl)benzene |
Specifications
| 589-15-1 | |
| 100.0 | |
| Tan to White | |
| Authentic | |
| Glass bottle | |
| BrC6H4CH2Br | |
| 25 g | |
| 15, 1416 | |
| Solubility in water: may decompose. Other solubilities: soluble in glacial acetic acid,soluble in alcohol,ether,carbon disulfide and,benzene | |
| C1=CC(=CC=C1CBr)Br | |
| 249.93 | |
| 249.93 | |
| Coarse Crystalline Solid or Solid Melt |
| 97.5 | |
| 60.0°C to 64.0°C | |
| 115.0°C to 124.0°C (12.0 mmHg) | |
| 97.5% min. (GC) | |
| C7H6Br2 | |
| MFCD00000179 | |
| 05, 308 | |
| 4-bromobenzyl bromide, 1-bromo-4-bromomethyl benzene, p-bromobenzyl bromide, alpha,p-dibromotoluene, alpha,4-dibromotoluene, benzene, 1-bromo-4-bromomethyl, 4-bromobenzylbromide, unii-3d1ttl8bgn, a,p-dibromotoluene, 4-bromo benzyl bromide | |
| YLRBJYMANQKEAW-UHFFFAOYSA-N | |
| 1-bromo-4-(bromomethyl)benzene | |
| 68527 | |
| 98% | |
| 4-Bromobenzyl bromide, 98% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
EINECSNumber : 209-636-5
TSCA : TSCA
RUO – Research Use Only