missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromobenzoyl Chloride 98.0+%, TCI America™
Supplier: TCI America B055825G
Specifications
| 4-Bromobenzoyl Chloride | |
| 38°C | |
| C7H4BrClO | |
| 25 g | |
| p-bromobenzoyl chloride, benzoyl chloride, 4-bromo, benzoyl chloride, p-bromo, 4-bromobenzoylchloride, p-bromobenzoyl chloride, p-bromo, 4-bromobenzoic acid chloride, p-brombenzoylchlorid, 4bromobenzoyl chloride, p-brc6h4cocl, 4-bromo-benzoylchloride | |
| ClC(=O)C1=CC=C(Br)C=C1 | |
| 219.46 | |
| 219.46 | |
| Crystalline Powder |
| 586-75-4 | |
| 247°C | |
| MFCD00000683 | |
| 3261 | |
| DENKGPBHLYFNGK-UHFFFAOYSA-N | |
| 4-bromobenzoyl chloride | |
| 68515 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 586-75-4 | |
| 219.46 | |
| DENKGPBHLYFNGK-UHFFFAOYSA-N | |
| 68515 | |
| ClC(=O)C1=CC=C(Br)C=C1 |
| C7H4BrClO | |
| MFCD00000683 | |
| p-bromobenzoyl chloride, benzoyl chloride, 4-bromo, benzoyl chloride, p-bromo, 4-bromobenzoylchloride, p-bromobenzoyl chloride, p-bromo, 4-bromobenzoic acid chloride, p-brombenzoylchlorid, 4bromobenzoyl chloride, p-brc6h4cocl, 4-bromo-benzoylchloride | |
| 4-bromobenzoyl chloride |
Safety and Handling
EINECSNumber : (3)-0062
TSCA : Yes