accessibility menu, dialog, popup

UserName

ClOBr

CAS RN 586-75-4

IUPAC Name: 4-bromobenzoyl chloride
Synonyms: benzoyl chloride, 4-bromo-
Molecular Weight (g/mol): 219.46
Molecular Formula: C7H4BrClO
InChi Key: DENKGPBHLYFNGK-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC=C(Br)C=C1
Boiling Point 
  • (2)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)