missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzoyl chloride, 98+%

Catalog No. AAL0379418
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0379418 50 g
AAL0379409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0379418 Supplier Thermo Scientific Chemicals Supplier No. L0379418
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 586-75-4 | C7H4BrClO | 219.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
4-Bromobenzoyl chloride is used in the synthesis of HIV-1 protease inhibitors. It is also used in the synthesis of oxazoles, cyclohexanone derivatives as analgesic and antimicrobial agents.

Solubility
Soluble in methanol. Reacts with water forming Hcl.

Notes
Moisture Sensitive. Store away from strong bases. Incompatible with moisture and bases. Store at 2 - 8°C.

Chemical Identifiers

CAS 586-75-4
Molecular Formula C7H4BrClO
Molecular Weight (g/mol) 219.46
MDL Number MFCD00000683
InChI Key DENKGPBHLYFNGK-UHFFFAOYSA-N
Synonym p-bromobenzoyl chloride, benzoyl chloride, 4-bromo, benzoyl chloride, p-bromo, 4-bromobenzoylchloride, p-bromobenzoyl chloride, p-bromo, 4-bromobenzoic acid chloride, p-brombenzoylchlorid, 4bromobenzoyl chloride, p-brc6h4cocl, 4-bromo-benzoylchloride
PubChem CID 68515
IUPAC Name 4-bromobenzoyl chloride
SMILES ClC(=O)C1=CC=C(Br)C=C1

Specifications

Melting Point 38°C to 42°C
Boiling Point 246°C
Flash Point >110°C (230°F)
Quantity 50 g
UN Number UN3261
Beilstein 636641
Sensitivity Moisture sensitive
Solubility Information Soluble in methanol. Reacts with water forming Hcl.
Formula Weight 219.47
Percent Purity ≥98%
Chemical Name or Material 4-Bromobenzoyl chloride
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 209-580-1
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only