missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromobenzonitrile 97.0+%, TCI America™
Supplier: TCI America B142625G
Specifications
| 4-Bromobenzonitrile | |
| 113°C | |
| 236°C | |
| MFCD00001811 | |
| 3439 | |
| HQSCPPCMBMFJJN-UHFFFAOYSA-N | |
| 4-bromobenzonitrile | |
| 12162 | |
| ≥97.0% (GC) |
| 623-00-7 | |
| White-Yellow | |
| C7H4BrN | |
| 25 g | |
| p-bromobenzonitrile, benzonitrile, 4-bromo, benzonitrile, p-bromo, 1-bromo-4-cyanobenzene, 4-bromo benzonitrile, 4-cyano-bromobenzene, 4-bromo-4'-cyanobenzene, 4-cyanobromobenzene, bromobenzonitrile-4, 4-bromocyanobenzene | |
| BrC1=CC=C(C=C1)C#N | |
| 182.02 | |
| 182.02 | |
| Crystalline Powder |
Chemical Identifiers
| 623-00-7 | |
| 182.02 | |
| HQSCPPCMBMFJJN-UHFFFAOYSA-N | |
| 12162 | |
| BrC1=CC=C(C=C1)C#N |
| C7H4BrN | |
| MFCD00001811 | |
| p-bromobenzonitrile, benzonitrile, 4-bromo, benzonitrile, p-bromo, 1-bromo-4-cyanobenzene, 4-bromo benzonitrile, 4-cyano-bromobenzene, 4-bromo-4'-cyanobenzene, 4-cyanobromobenzene, bromobenzonitrile-4, 4-bromocyanobenzene | |
| 4-bromobenzonitrile |
Safety and Handling
RTECSNumber : DI2460000
TSCA : No