missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzonitrile 97.0+%, TCI America™

Catalog No. p-7119048
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B142625G 25 g
B1426250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B142625G Supplier TCI America Supplier No. B142625G
Only null left

Chemical Identifiers

CAS 623-00-7
Molecular Formula C7H4BrN
Molecular Weight (g/mol) 182.02
MDL Number MFCD00001811
InChI Key HQSCPPCMBMFJJN-UHFFFAOYSA-N
Synonym p-bromobenzonitrile, benzonitrile, 4-bromo, benzonitrile, p-bromo, 1-bromo-4-cyanobenzene, 4-bromo benzonitrile, 4-cyano-bromobenzene, 4-bromo-4'-cyanobenzene, 4-cyanobromobenzene, bromobenzonitrile-4, 4-bromocyanobenzene
PubChem CID 12162
IUPAC Name 4-bromobenzonitrile
SMILES BrC1=CC=C(C=C1)C#N

Specifications

Melting Point 113°C
Color White-Yellow
Boiling Point 236°C
Quantity 25 g
UN Number 3439
Formula Weight 182.02
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromobenzonitrile
RTECSNumber DI2460000
TSCA No