Learn More
4-Bromobenzoic acid, 97%
CAS: 586-76-5 | C7H5BrO2 | 201.02 g/mol
Supplier: Thermo Scientific Chemicals A1251822
Description
4-Bromobenzoic acid was used to study the metabolic fate of 2-,3-and 4-bromo benzoic acids in rat hepatocytes incubation using high temperature liquid chromatography. It was used in bromine-specific detection of the metabolites of 2-,3-and 4-bromobenzoic acid in the urine and bile of rats by inductively coupled plasma mass spectrometry. It is also used as an intermediate for the synthesis of agrochemicals, pharmaceutical, chemical intermediates and liquid crystals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Bromobenzoic acid was used to study the metabolic fate of 2-,3-and 4-bromo benzoic acids in rat hepatocytes incubation using high temperature liquid chromatography. It was used in bromine-specific detection of the metabolites of 2-,3-and 4-bromobenzoic acid in the urine and bile of rats by inductively coupled plasma mass spectrometry. It is also used as an intermediate for the synthesis of agrochemicals, pharmaceutical, chemical intermediates and liquid crystals.
Solubility
Slightly soluble in hot water. Soluble in alcohol and ether.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
| 4-Bromobenzoic acid | |
| 253°C to 258°C | |
| Odorless | |
| MFCD00002529 | |
| 1906923 | |
| p-bromobenzoic acid, benzoic acid, 4-bromo, p-carboxybromobenzene, benzoic acid, p-bromo, p-bromobenzenecarboxylic acid, 4-bromo-benzoic acid, 4-brom-benzoesaeure, 4-carboxybromobenzene, para bromo benzoic acid, 4bromo-benzoic acid | |
| TUXYZHVUPGXXQG-UHFFFAOYSA-N | |
| 4-bromobenzoic acid | |
| 11464 | |
| 201.02 |
| 586-76-5 | |
| 1.894 | |
| C7H5BrO2 | |
| 100 g | |
| 14,1408 | |
| Slightly soluble in hot water. Soluble in alcohol and ether. | |
| OC(=O)C1=CC=C(Br)C=C1 | |
| 201.02 | |
| CHEBI:60698 | |
| ≥98% |
Chemical Identifiers
| 586-76-5 | |
| 201.02 | |
| TUXYZHVUPGXXQG-UHFFFAOYSA-N | |
| 11464 | |
| 4-bromobenzoic acid |
| C7H5BrO2 | |
| MFCD00002529 | |
| p-bromobenzoic acid, benzoic acid, 4-bromo, p-carboxybromobenzene, benzoic acid, p-bromo, p-bromobenzenecarboxylic acid, 4-bromo-benzoic acid, 4-brom-benzoesaeure, 4-carboxybromobenzene, para bromo benzoic acid, 4bromo-benzoic acid | |
| CHEBI:60698 | |
| OC(=O)C1=CC=C(Br)C=C1 |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 209-581-7
RTECSNumber : DG4448050
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only