accessibility menu, dialog, popup

UserName

HOOBr

CAS RN 586-76-5

IUPAC Name: 4-bromobenzoic acid
Synonyms: 4-bromobenzoic acid 4-brom-benzoesaeure P-bromobenzoic acid
Molecular Weight (g/mol): 201.02
Molecular Formula: C7H5BrO2
InChi Key: TUXYZHVUPGXXQG-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(Br)C=C1
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (3)
  • (3)
  • (2)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)