missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromobenzohydrazide 97.0+%, TCI America™
Supplier: TCI America B206725G
Specifications
| 4-Bromobenzohydrazide | |
| 168°C | |
| C7H7BrN2O | |
| 25 g | |
| UYIMBYKIIMYFPS-UHFFFAOYSA-N | |
| 4-bromobenzohydrazide | |
| 22219 | |
| ≥97.0% (T) |
| 5933-32-4 | |
| White | |
| MFCD00007602 | |
| 4-bromobenzhydrazide, p-bromobenzhydrazide, 4-bromobenzoic hydrazide, p-bromobenzohydrazide, p-bromobenzoic acid hydrazide, 4-bromobenzoylhydrazine, 4-bromobenzoic acid hydrazide, benzoic acid, 4-bromo-, hydrazide, 4-bromo-benzoic acid hydrazide, p-bromobenzoylhydrazine | |
| C1=CC(=CC=C1C(=O)NN)Br | |
| 215.05 | |
| 215.05 | |
| Crystalline Powder |
Chemical Identifiers
| 5933-32-4 | |
| 215.05 | |
| UYIMBYKIIMYFPS-UHFFFAOYSA-N | |
| 22219 | |
| C1=CC(=CC=C1C(=O)NN)Br |
| C7H7BrN2O | |
| MFCD00007602 | |
| 4-bromobenzhydrazide, p-bromobenzhydrazide, 4-bromobenzoic hydrazide, p-bromobenzohydrazide, p-bromobenzoic acid hydrazide, 4-bromobenzoylhydrazine, 4-bromobenzoic acid hydrazide, benzoic acid, 4-bromo-, hydrazide, 4-bromo-benzoic acid hydrazide, p-bromobenzoylhydrazine | |
| 4-bromobenzohydrazide |
Safety and Handling
RTECSNumber : DG4449250
TSCA : No