missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzeneboronic acid neopentyl glycol ester, 98+%

CAS: 183677-71-6 | C11H14BBrO2 | 268.945 g/mol

Supplier:  Thermo Scientific Chemicals L1745503

 View more versions of this product

Catalog No. AAL1745503


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Bromobenzeneboronic acid neopentyl glycol ester
109°C to 111°C
MFCD01318105
2-4-bromophenyl-5,5-dimethyl-1,3,2-dioxaborinane, 4-bromobenzeneboronic acid neopentyl glycol ester, 4-bromophenylboronic acid neopentyl glycol ester, amtb980, 2-4-bromophenyl-5,5-dimethyl-2-bora-1,3-dioxane, 4-bromophenylboronic acid neopentyl glycol cyclic ester
B1(OCC(CO1)(C)C)C2=CC=C(C=C2)Br
268.945
268.95
183677-71-6
C11H14BBrO2
1 g
GUASPWAWPYERCR-UHFFFAOYSA-N
2-(4-bromophenyl)-5,5-dimethyl-1,3,2-dioxaborinane
10400865
≥98%

Chemical Identifiers

183677-71-6
268.945
GUASPWAWPYERCR-UHFFFAOYSA-N
10400865
B1(OCC(CO1)(C)C)C2=CC=C(C=C2)Br
C11H14BBrO2
MFCD01318105
2-4-bromophenyl-5,5-dimethyl-1,3,2-dioxaborinane, 4-bromobenzeneboronic acid neopentyl glycol ester, 4-bromophenylboronic acid neopentyl glycol ester, amtb980, 2-4-bromophenyl-5,5-dimethyl-2-bora-1,3-dioxane, 4-bromophenylboronic acid neopentyl glycol cyclic ester
2-(4-bromophenyl)-5,5-dimethyl-1,3,2-dioxaborinane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only