accessibility menu, dialog, popup

UserName

BOOH3CCH3Br

CAS RN 183677-71-6

IUPAC Name: 2-(4-bromophenyl)-5,5-dimethyl-1,3,2-dioxaborinane
Synonyms: 2-(4-bromophenyl)-5,5-dimethyl-1,3,2-dioxaborinane
Molecular Weight (g/mol): 268.95
Molecular Formula: C11H14BBrO2
InChi Key: GUASPWAWPYERCR-UHFFFAOYSA-N
SMILES: CC1(C)COB(OC1)C1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results