missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzeneboronic acid neopentyl glycol ester, 98+%

Catalog No. AAL1745506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1745506 5 g
AAL1745503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1745506 Supplier Thermo Scientific Chemicals Supplier No. L1745506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 183677-71-6 | C11H14BBrO2 | 268.945 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 183677-71-6
Molecular Formula C11H14BBrO2
Molecular Weight (g/mol) 268.945
MDL Number MFCD01318105
InChI Key GUASPWAWPYERCR-UHFFFAOYSA-N
Synonym 2-4-bromophenyl-5,5-dimethyl-1,3,2-dioxaborinane, 4-bromobenzeneboronic acid neopentyl glycol ester, 4-bromophenylboronic acid neopentyl glycol ester, amtb980, 2-4-bromophenyl-5,5-dimethyl-2-bora-1,3-dioxane, 4-bromophenylboronic acid neopentyl glycol cyclic ester
PubChem CID 10400865
IUPAC Name 2-(4-bromophenyl)-5,5-dimethyl-1,3,2-dioxaborinane
SMILES B1(OCC(CO1)(C)C)C2=CC=C(C=C2)Br

Specifications

Melting Point 109°C to 111°C
Quantity 5 g
Formula Weight 268.95
Percent Purity ≥98%
Chemical Name or Material 4-Bromobenzeneboronic acid neopentyl glycol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only