missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzamidine Hydrochloride 98.0+%, TCI America™

Catalog No. B41611G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41611G 1 g
B41615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41611G Supplier TCI America Supplier No. B41611G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 55368-42-8
Molecular Formula C7H8BrClN2
Molecular Weight (g/mol) 235.509
MDL Number MFCD00464967
InChI Key IMTHEBSPHHMJOJ-UHFFFAOYSA-N
Synonym 4-bromobenzamidine hydrochloride, 4-bromobenzimidamide hydrochloride, 4-bromobenzamidine hcl, 4-bromo-benzamidine hydrochloride, 4-bromobenzenecarboximidamide hydrochloride, 4-bromobenzene-1-carboximidamide hydrochloride, benzenecarboximidamide, 4-bromo-, monohydrochloride, 4-bromobenzenecarboxamidine, chloride, benzenecarboximidamide,4-bromo, 4-bromobenzamidinehydrochloride
PubChem CID 12207714
IUPAC Name 4-bromobenzenecarboximidamide;hydrochloride
SMILES C1=CC(=CC=C1C(=N)N)Br.Cl

Specifications

Melting Point 265°C
Color White
Quantity 1 g
Formula Weight 235.51
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromobenzamidine Hydrochloride