accessibility menu, dialog, popup

UserName

CAS RN 55368-42-8

IUPAC Name: 4-bromobenzene-1-carboximidamide hydrochloride
Synonyms: 4-bromobenzenecarboximidamide hydrochloride
Molecular Weight (g/mol): 235.51
Molecular Formula: C7H8BrClN2
InChi Key: IMTHEBSPHHMJOJ-UHFFFAOYSA-N
SMILES: Cl.NC(=N)C1=CC=C(Br)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results