missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromobenzamide 98.0+%, TCI America™
$70.84 - $281.48
Chemical Identifiers
| CAS | 698-67-9 |
|---|---|
| Molecular Formula | C7H6BrNO |
| Molecular Weight (g/mol) | 200.035 |
| MDL Number | MFCD00007991 |
| InChI Key | ZRWNRAJCPNLYAK-UHFFFAOYSA-N |
| Synonym | p-bromobenzamide, benzamide, 4-bromo, benzamide, p-bromo, p-bromobenzoic acid amide, 4-bromo-benzamide, 4-bromo benzamide, pubchem3737, acmc-209obe, 4-bromobenzamide, 4-bromobenzoic acid amide |
| PubChem CID | 69683 |
| IUPAC Name | 4-bromobenzamide |
| SMILES | C1=CC(=CC=C1C(=O)N)Br |
Chemical Identifiers
| 698-67-9 | |
| 200.035 | |
| ZRWNRAJCPNLYAK-UHFFFAOYSA-N | |
| 69683 | |
| C1=CC(=CC=C1C(=O)N)Br |
| C7H6BrNO | |
| MFCD00007991 | |
| p-bromobenzamide, benzamide, 4-bromo, benzamide, p-bromo, p-bromobenzoic acid amide, 4-bromo-benzamide, 4-bromo benzamide, pubchem3737, acmc-209obe, 4-bromobenzamide, 4-bromobenzoic acid amide | |
| 4-bromobenzamide |
Specifications
| 698-67-9 | |
| White-Yellow | |
| MFCD00007991 | |
| p-bromobenzamide, benzamide, 4-bromo, benzamide, p-bromo, p-bromobenzoic acid amide, 4-bromo-benzamide, 4-bromo benzamide, pubchem3737, acmc-209obe, 4-bromobenzamide, 4-bromobenzoic acid amide | |
| C1=CC(=CC=C1C(=O)N)Br | |
| 200.035 | |
| 200.04 | |
| Crystalline Powder |
| 193°C | |
| C7H6BrNO | |
| 5 g | |
| ZRWNRAJCPNLYAK-UHFFFAOYSA-N | |
| 4-bromobenzamide | |
| 69683 | |
| ≥98.0% (HPLC,N) | |
| 4-Bromobenzamide |
Safety and Handling
RTECSNumber : CV2365800
TSCA : No