missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromobenzaldehyde 97.0+%, TCI America™
Supplier: TCI America B0549250G
Specifications
| 4-Bromobenzaldehyde | |
| 57°C | |
| 125°C | |
| MFCD00003377 | |
| p-bromobenzaldehyde, benzaldehyde, 4-bromo, benzaldehyde, p-bromo, 4-bromo-benzaldehyde, 4-bromo benzaldehyde, 4-bromobenzylaldehyde, p-bromo benzaldehyde, unii-4l8vm24f65, 4-brombenzaldehyde, 4-brombenzaldehyd | |
| C1=CC(=CC=C1C=O)Br | |
| 185.02 | |
| 185.02 | |
| Crystalline Powder |
| 1122-91-4 | |
| White-Yellow | |
| C7H5BrO | |
| 250 g | |
| ZRYZBQLXDKPBDU-UHFFFAOYSA-N | |
| 4-bromobenzaldehyde | |
| 70741 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 1122-91-4 | |
| 185.02 | |
| ZRYZBQLXDKPBDU-UHFFFAOYSA-N | |
| 70741 | |
| C1=CC(=CC=C1C=O)Br |
| C7H5BrO | |
| MFCD00003377 | |
| p-bromobenzaldehyde, benzaldehyde, 4-bromo, benzaldehyde, p-bromo, 4-bromo-benzaldehyde, 4-bromo benzaldehyde, 4-bromobenzylaldehyde, p-bromo benzaldehyde, unii-4l8vm24f65, 4-brombenzaldehyde, 4-brombenzaldehyd | |
| 4-bromobenzaldehyde |
Safety and Handling
RTECSNumber : CU4810000
TSCA : Yes