accessibility menu, dialog, popup

UserName

BrO

CAS RN 1122-91-4

IUPAC Name: 4-bromobenzaldehyde
Synonyms: benzaldehyde, 4-bromo-
Molecular Weight (g/mol): 185.02
Molecular Formula: C7H5BrO
InChi Key: ZRYZBQLXDKPBDU-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=O)C=C1
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (2)
  • (3)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)