missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzaldehyde Diethyl Acetal 97.0+%, TCI America™

Catalog No. B284525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B284525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B284525G Supplier TCI America Supplier No. B284525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 34421-94-8
Molecular Formula C11H15BrO2
Molecular Weight (g/mol) 259.14
MDL Number MFCD01863514
InChI Key BFSNEBVTOODGHZ-UHFFFAOYSA-N
Synonym 4-bromobenzaldehyde diethyl acetal, 1-bromo-4-diethoxymethyl benzene, 4-bromobenzaldehydediethylacetal, benzene, 1-bromo-4-diethoxymethyl, 4-bromophenyl diethoxymethane, 4-bromobenzaldehyde diethylacetal, 4-bromo-alpha,alpha-diethoxytoluene, pubchem5372, acmc-1aigt, 4-bromobenzaldehydediethyl acetal
PubChem CID 688340
IUPAC Name 1-bromo-4-(diethoxymethyl)benzene
SMILES CCOC(OCC)C1=CC=C(Br)C=C1

Specifications

Color Yellow
Quantity 25 g
Formula Weight 259.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromobenzaldehyde Diethyl Acetal