accessibility menu, dialog, popup

UserName

CAS RN 34421-94-8

IUPAC Name: 1-bromo-4-(diethoxymethyl)benzene
Synonyms: 1-bromo-4-(diethoxymethyl)benzene
Molecular Weight (g/mol): 259.14
Molecular Formula: C11H15BrO2
InChi Key: BFSNEBVTOODGHZ-UHFFFAOYSA-N
SMILES: CCOC(OCC)C1=CC=C(Br)C=C1
Percent Purity 
  • (1)
  • (1)
  • (1)
Quantity 
  • (1)
  • (2)
Boiling Point 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)