missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzaldehyde, 98+%

CAS: 1122-91-4 | C7H5BrO | 185.02 g/mol

$161.36 - $1737.05

Chemical Identifiers

CAS 1122-91-4
Molecular Formula C7H5BrO
Molecular Weight (g/mol) 185.02
MDL Number MFCD00003377
InChI Key ZRYZBQLXDKPBDU-UHFFFAOYSA-N
Synonym p-bromobenzaldehyde, benzaldehyde, 4-bromo, benzaldehyde, p-bromo, 4-bromo-benzaldehyde, 4-bromo benzaldehyde, 4-bromobenzylaldehyde, p-bromo benzaldehyde, unii-4l8vm24f65, 4-brombenzaldehyde, 4-brombenzaldehyd
PubChem CID 70741
IUPAC Name 4-bromobenzaldehyde
SMILES C1=CC(=CC=C1C=O)Br
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1423714
View Documents
Thermo Scientific Chemicals
A1423714
25 g
Each for $161.36
Only null left
 
AAA1423722
View Documents
Thermo Scientific Chemicals
A1423722
100 g
Each for $446.62
Only null left
 
AAA1423736
View Documents
Thermo Scientific Chemicals
A1423736
500 g
Each for $1,737.05
Only null left
 
Description

Description

4-Bromobenzaldehyde is widely utilized as an intermediate for the preparation of agrochemicals, pharmaceuticals and other organic compounds. It is used as a precursor and reacts with sulfamethoxazole to prepare Schiff’s base, 4-[(4-Bromo-benzylidene)-amino]-N-(5-methyl-isoxazol-3-yl)-benzene sulfonamide, which can be used for the photo stability of polyvinyl chloride (PVC). It plays an important role in palladium-catalyzed mono- and bis-arylation of 3,4-(ethylenedioxy)thiophenes. It is employed in a cross-coupling study with potassium vinyltrifluoroborate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobenzaldehyde is widely utilized as an intermediate for the preparation of agrochemicals, pharmaceuticals and other organic compounds. It is used as a precursor and reacts with sulfamethoxazole to prepare Schiff’s base, 4-[(4-Bromo-benzylidene)-amino]-N-(5-methyl-isoxazol-3-yl)-benzene sulfonamide, which can be used for the photo stability of polyvinyl chloride (PVC). It plays an important role in palladium-catalyzed mono- and bis-arylation of 3,4-(ethylenedioxy)thiophenes. It is employed in a cross-coupling study with potassium vinyltrifluoroborate.

Solubility
Soluble in ethanol and benzene. Insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong bases, strong oxidizing agents and strong reducing agents.
Specifications

Chemical Identifiers

1122-91-4
185.02
ZRYZBQLXDKPBDU-UHFFFAOYSA-N
70741
C1=CC(=CC=C1C=O)Br
C7H5BrO
MFCD00003377
p-bromobenzaldehyde, benzaldehyde, 4-bromo, benzaldehyde, p-bromo, 4-bromo-benzaldehyde, 4-bromo benzaldehyde, 4-bromobenzylaldehyde, p-bromo benzaldehyde, unii-4l8vm24f65, 4-brombenzaldehyde, 4-brombenzaldehyd
4-bromobenzaldehyde

Specifications

1122-91-4
66°C to 68°C (2 mmHg)
Characteristic
MFCD00003377
507100
p-bromobenzaldehyde, benzaldehyde, 4-bromo, benzaldehyde, p-bromo, 4-bromo-benzaldehyde, 4-bromo benzaldehyde, 4-bromobenzylaldehyde, p-bromo benzaldehyde, unii-4l8vm24f65, 4-brombenzaldehyde, 4-brombenzaldehyd
ZRYZBQLXDKPBDU-UHFFFAOYSA-N
4-bromobenzaldehyde
70741
≥98%
55°C to 58°C
108°C (226°F)
C7H5BrO
25 g
Air Sensitive
Soluble in ethanol and benzene. Insoluble in water.
C1=CC(=CC=C1C=O)Br
185.02
185.03
4-Bromobenzaldehyde
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P342+P311-P362-P501c

H302-H315-H319-H334-H335

EINECSNumber : 214-365-0

RTECSNumber : CU4810000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only