missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenz[a]anthracene 97.0+%, TCI America™

Catalog No. B5450200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5450200MG 200 mg
B54501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5450200MG Supplier TCI America Supplier No. B5450200MG
Only null left

Chemical Identifiers

CAS 61921-39-9
Molecular Formula C18H11Br
Molecular Weight (g/mol) 307.19
MDL Number MFCD25371993
InChI Key ILVKEMDEUCXCAB-UHFFFAOYSA-N
Synonym 4-Bromotetraphene
PubChem CID 182324
IUPAC Name 4-bromobenzo[a]anthracene
SMILES C1=CC=C2C=C3C(=CC2=C1)C=CC4=C3C=CC=C4Br

Specifications

Melting Point 220°C
Quantity 200 mg
Formula Weight 307.19
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromobenz[a]anthracene