accessibility menu, dialog, popup

UserName

Br

CAS RN 61921-39-9

IUPAC Name: 4-bromotetraphene
Synonyms: 4-bromotetraphene
Molecular Weight (g/mol): 307.19
Molecular Formula: C18H11Br
InChi Key: ILVKEMDEUCXCAB-UHFFFAOYSA-N
SMILES: BrC1=C2C=CC3=CC4=CC=CC=C4C=C3C2=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results