missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Bromoacetophenone 98.0+%, TCI America™
$63.52 - $463.97
Chemical Identifiers
| CAS | 99-90-1 |
|---|---|
| Molecular Formula | C8H7BrO |
| Molecular Weight (g/mol) | 199.05 |
| MDL Number | MFCD00000105 |
| InChI Key | WYECURVXVYPVAT-UHFFFAOYSA-N |
| Synonym | 4'-bromoacetophenone, 1-4-bromophenyl ethanone, p-bromoacetophenone, 4-bromoacetophenone, ethanone, 1-4-bromophenyl, 1-4-bromophenyl ethan-1-one, 1-acetyl-4-bromobenzene, methyl p-bromophenyl ketone, p-bromophenyl methyl ketone, acetophenone, 4'-bromo |
| PubChem CID | 7466 |
| IUPAC Name | 1-(4-bromophenyl)ethan-1-one |
| SMILES | CC(=O)C1=CC=C(Br)C=C1 |
Chemical Identifiers
| 99-90-1 | |
| 199.05 | |
| WYECURVXVYPVAT-UHFFFAOYSA-N | |
| 7466 | |
| CC(=O)C1=CC=C(Br)C=C1 |
| C8H7BrO | |
| MFCD00000105 | |
| 4'-bromoacetophenone, 1-4-bromophenyl ethanone, p-bromoacetophenone, 4-bromoacetophenone, ethanone, 1-4-bromophenyl, 1-4-bromophenyl ethan-1-one, 1-acetyl-4-bromobenzene, methyl p-bromophenyl ketone, p-bromophenyl methyl ketone, acetophenone, 4'-bromo | |
| 1-(4-bromophenyl)ethan-1-one |
Specifications
| 99-90-1 | |
| 256°C | |
| MFCD00000105 | |
| 4'-bromoacetophenone, 1-4-bromophenyl ethanone, p-bromoacetophenone, 4-bromoacetophenone, ethanone, 1-4-bromophenyl, 1-4-bromophenyl ethan-1-one, 1-acetyl-4-bromobenzene, methyl p-bromophenyl ketone, p-bromophenyl methyl ketone, acetophenone, 4'-bromo | |
| CC(=O)C1=CC=C(Br)C=C1 | |
| 199.05 | |
| 199.05 | |
| Crystalline Lumps |
| 53°C | |
| C8H7BrO | |
| 25 g | |
| WYECURVXVYPVAT-UHFFFAOYSA-N | |
| 1-(4-bromophenyl)ethan-1-one | |
| 7466 | |
| ≥98.0% (GC) | |
| 4′-Bromoacetophenone |
Safety and Handling
TSCA : Yes