accessibility menu, dialog, popup

UserName

CAS RN 99-90-1

IUPAC Name: 1-(4-bromophenyl)ethan-1-one
Synonyms: P-bromoacetophenone
Molecular Weight (g/mol): 199.05
Molecular Formula: C8H7BrO
InChi Key: WYECURVXVYPVAT-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(Br)C=C1
Boiling Point 
  • (3)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (2)
  • (3)
  • (3)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (3)
  • (2)
  • (2)
  • (6)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)