missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromoacetophenone 98.0+%, TCI America™

Catalog No. b053725g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
B053725G 25 g
B0537500G 500 g
2 options

Catalog No. B053725G

Supplier: TCI America B053725G

Only null left

Chemical Identifiers

CAS 99-90-1
Molecular Formula C8H7BrO
Molecular Weight (g/mol) 199.05
MDL Number MFCD00000105
InChI Key WYECURVXVYPVAT-UHFFFAOYSA-N
Synonym 4'-bromoacetophenone, 1-4-bromophenyl ethanone, p-bromoacetophenone, 4-bromoacetophenone, ethanone, 1-4-bromophenyl, 1-4-bromophenyl ethan-1-one, 1-acetyl-4-bromobenzene, methyl p-bromophenyl ketone, p-bromophenyl methyl ketone, acetophenone, 4'-bromo
PubChem CID 7466
IUPAC Name 1-(4-bromophenyl)ethan-1-one
SMILES CC(=O)C1=CC=C(Br)C=C1

Specifications

Melting Point 53°C
Boiling Point 256°C
Quantity 25 g
Formula Weight 199.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4′-Bromoacetophenone