missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-o-phenylenediamine, 97%

Catalog No. AAH2692906
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2692906 5 g
AAH2692914 25 g
AAH2692922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2692906 Supplier Thermo Scientific Chemicals Supplier No. H2692906

CAS: 1575-37-7 | C6H7BrN2 | 187.04 g/mol

4-Bromo-o-phenylenediamine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-o-phenylenediamine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.

Chemical Identifiers

CAS 1575-37-7
Molecular Formula C6H7BrN2
Molecular Weight (g/mol) 187.04
MDL Number MFCD02660622
InChI Key WIHHVKUARKTSBU-UHFFFAOYSA-N
Synonym 4-bromo-1,2-benzenediamine, 4-bromo-1,2-diaminobenzene, 4-bromo-o-phenylenediamine, 1,2-diamino-4-bromobenzene, 2-amino-4-bromoaniline, 4-bromo-1,2-phenylenediamine, 4-bromo-2-aminoaniline, 1,2-benzenediamine, 4-bromo, 2-amino-4-bromophenylamine, 4-bromophenylene-1,2-diamine
PubChem CID 323593
IUPAC Name 4-bromobenzene-1,2-diamine
SMILES NC1=CC=C(Br)C=C1N

Specifications

Melting Point 64°C to 66°C
Quantity 5 g
UN Number UN2811
Solubility Information Soluble in chloroform.
Formula Weight 187.04
Percent Purity 97%
Chemical Name or Material 4-Bromo-o-phenylenediamine
Hazard Category H301-H315-H317-H319
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P272-P280-P301+P310-P302+P352-P305+P351+P338-P330-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only