missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-N,N,3,5-tetramethylaniline 98.0+%, TCI America™

Catalog No. p-7119927
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30701G 1 g
B30705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30701G Supplier TCI America Supplier No. B30701G
Only null left

Chemical Identifiers

CAS 14275-09-3
Molecular Formula C10H14BrN
Molecular Weight (g/mol) 228.13
MDL Number MFCD11036245
InChI Key AQJHDBKXJXXDKV-UHFFFAOYSA-N
Synonym 2-Bromo-5-(dimethylamino)-m-xylene, 4-Bromo-N,N-dimethyl-3,5-xylidine
PubChem CID 44629944
IUPAC Name 4-bromo-N,N,3,5-tetramethylaniline
SMILES CN(C)C1=CC(C)=C(Br)C(C)=C1

Specifications

Color White-Yellow
Quantity 1 g
Physical Form Crystal-Powder at 20°C
Formula Weight 228.13
Percent Purity ≥98.0% (GC)
Chemical Name or Material 4-Bromo-N,N,3,5-tetramethylaniline
TSCA No
Recommended Storage Refrigerator