accessibility menu, dialog, popup

UserName

CAS RN 14275-09-3

CH3BrH3CNCH3CH3

CAS RN 14275-09-3

IUPAC Name: 4-bromo-N,N,3,5-tetramethylaniline
Synonyms: 4-bromo-N,N,3,5-tetramethylaniline
Molecular Weight (g/mol): 228.13
Molecular Formula: C10H14BrN
InChi Key: AQJHDBKXJXXDKV-UHFFFAOYSA-N
SMILES: CN(C)C1=CC(C)=C(Br)C(C)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results