missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-N-methylbenzylamine, 97%

Catalog No. p-7036245
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5450603 1 g
AAH5450606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5450603 Supplier Thermo Scientific Chemicals Supplier No. H5450603
Only null left

CAS: 699-03-6 | C8H10BrN | 200.079 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 699-03-6
Molecular Formula C8H10BrN
Molecular Weight (g/mol) 200.079
MDL Number MFCD04507522
InChI Key URFJXIULELMVHV-UHFFFAOYSA-N
Synonym n-methyl-4-bromobenzylamine, 1-4-bromophenyl-n-methylmethanamine, 4-bromo-n-methylbenzylamine, 4-bromobenzyl methylamine, benzenemethanamine, 4-bromo-n-methyl, 4-bromophenyl methyl methyl amine, 4-bromo methylbenzylamine, 1-4-bromophenyl-n-methyl-methanamine, acmc-20a2f7, 4-bromo-benzyl-methyl-amine
PubChem CID 485400
IUPAC Name 1-(4-bromophenyl)-N-methylmethanamine
SMILES CNCC1=CC=C(C=C1)Br

Specifications

Density 1.461
Boiling Point 231°C to 232°C
Flash Point >110°C (230°F)
Refractive Index 1.565
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 200.08
Percent Purity 97%
Chemical Name or Material 4-Bromo-N-methylbenzylamine
Hazard Category H302-H319
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only