accessibility menu, dialog, popup

UserName

CAS RN 699-03-6

IUPAC Name: [(4-bromophenyl)methyl](methyl)amine
Synonyms: [(4-bromophenyl)methyl](methyl)amine
Molecular Weight (g/mol): 200.08
Molecular Formula: C8H10BrN
InChi Key: URFJXIULELMVHV-UHFFFAOYSA-N
SMILES: CNCC1=CC=C(Br)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results