missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-N-methoxy-N-methylbenzamide 95.0+%, TCI America™

Catalog No. p-7124165
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B57281G 1 g
B57285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B57281G Supplier TCI America Supplier No. B57281G

Chemical Identifiers

CAS 192436-83-2
Molecular Formula C9H10BrNO2
Molecular Weight (g/mol) 244.088
MDL Number MFCD02684303
InChI Key YEHZWOBPONLEEH-UHFFFAOYSA-N
PubChem CID 10999252
IUPAC Name 4-bromo-N-methoxy-N-methylbenzamide
SMILES CN(C(=O)C1=CC=C(C=C1)Br)OC

Specifications

Color Yellow
Quantity 1 g
Formula Weight 244.09
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromo-N-methoxy-N-methylbenzamide