accessibility menu, dialog, popup

UserName

H3CNOBrOH3C

CAS RN 192436-83-2

IUPAC Name: 4-bromo-N-methoxy-N-methylbenzamide
Synonyms: 4-bromo-N-methoxy-N-methylbenzamide
Molecular Weight (g/mol): 244.09
Molecular Formula: C9H10BrNO2
InChi Key: YEHZWOBPONLEEH-UHFFFAOYSA-N
SMILES: CON(C)C(=O)C1=CC=C(Br)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results