missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-DL-phenylalanine, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 227792500

Quantity 250mg
Packaging Glass bottle

 View more versions of this product

Catalog No. AC227792500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

14091-15-7
244.09
PEMUHKUIQHFMTH-UHFFFAOYSA-N
85681
C1=CC(=CC=C1CC(C(=O)O)N)Br
C9H10BrNO2
MFCD00002599
2-amino-3-4-bromophenyl propanoic acid, p-bromo-dl-phenylalanine, 4-bromo-dl-phenylalanine, dl-4-br-phe-oh, 4-bromophenylalanine, 4-bromo-phenylalanine, h-dl-phe 4-br-oh, 2-amino-3-4-bromophenyl propionic acid, 2-amino-3-4-bromo-phenyl-propionic acid, p-br-phenylalanine
2-amino-3-(4-bromophenyl)propanoic acid

Specifications

4-Bromo-DL-phenylalanine
14091-15-7
100.0
99%
C9H10BrNO2
250mg
2-amino-3-4-bromophenyl propanoic acid, p-bromo-dl-phenylalanine, 4-bromo-dl-phenylalanine, dl-4-br-phe-oh, 4-bromophenylalanine, 4-bromo-phenylalanine, h-dl-phe 4-br-oh, 2-amino-3-4-bromophenyl propionic acid, 2-amino-3-4-bromo-phenyl-propionic acid, p-br-phenylalanine
C1=CC(=CC=C1CC(C(=O)O)N)Br
244.09
244.09
99%
98.5
Authentic
Glass bottle
BrC6H4CH2CH(NH2)CO2H
MFCD00002599
PEMUHKUIQHFMTH-UHFFFAOYSA-N
2-amino-3-(4-bromophenyl)propanoic acid
85681
99%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 237-941-3

SDS
Documents

Documents

Promotions

Promotions

Promo
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.

Adding...