accessibility menu, dialog, popup

UserName

NH2BrOHO

CAS RN 14091-15-7

IUPAC Name: 2-amino-3-(4-bromophenyl)propanoic acid
Synonyms: 2-amino-3-(4-bromophenyl)propanoic acid
Molecular Weight (g/mol): 244.09
Molecular Formula: C9H10BrNO2
InChi Key: PEMUHKUIQHFMTH-UHFFFAOYSA-N
SMILES: NC(CC1=CC=C(Br)C=C1)C(O)=O
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)